Product Details

SNP ID
rs140890109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23785476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTCTGAAGAGAGTTCAGAGTTA[C/T]AAATGCTTACAATGACTTTTTAAAT
Phenotype
MIM: 313020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAT1 PubMed Links

Gene Details

Gene
SAT1
Gene Name
spermidine/spermine N1-acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002970.3 Intron NP_002961.1
XM_017029720.1 Intron XP_016885209.1

View Full Product Details