Product Details
- SNP ID
-
rs35064500
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53795911 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCTCAGCCGCTTGCAAAGCAGTC[A/G]GACACCTGTGTCCCCTAGGGCGTTG
- Phenotype
-
MIM: 612043
MIM: 612044
MIM: 611954
MIM: 609648
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR371A
PubMed Links
Gene Details
- Gene
- MIR371A
- Gene Name
- microRNA 371a
There are no transcripts associated with this gene.
- Gene
- MIR371B
- Gene Name
- microRNA 371b
There are no transcripts associated with this gene.
- Gene
- MIR372
- Gene Name
- microRNA 372
There are no transcripts associated with this gene.
- Gene
- MIR373
- Gene Name
- microRNA 373
There are no transcripts associated with this gene.
- Gene
- NLRP12
- Gene Name
- NLR family pyrin domain containing 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001277126.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R1017* |
NP_001264055.1 |
NM_001277129.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R959* |
NP_001264058.1 |
NM_144687.3 |
3107 |
Nonsense Mutation |
CGA,TGA |
R1016* |
NP_653288.1 |
XM_011527479.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R960* |
XP_011525781.1 |
XM_011527480.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R960* |
XP_011525782.1 |
XM_011527482.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R903* |
XP_011525784.1 |
XM_017027460.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R1016* |
XP_016882949.1 |
XM_017027461.1 |
3107 |
Intron |
|
|
XP_016882950.1 |
XM_017027462.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R959* |
XP_016882951.1 |
XM_017027463.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R878* |
XP_016882952.1 |
XM_017027464.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R878* |
XP_016882953.1 |
XM_017027465.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R878* |
XP_016882954.1 |
XM_017027466.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R878* |
XP_016882955.1 |
XM_017027467.1 |
3107 |
Nonsense Mutation |
CGA,TGA |
R878* |
XP_016882956.1 |
View Full Product Details