Product Details

SNP ID
rs10172196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:36553406 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAATGAGATACCAAAAGGCCAT[A/G]TCTCTTAGGGTGCCACTTGGAGATA
Phenotype
MIM: 606189 MIM: 604826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRIM1 PubMed Links

Gene Details

Gene
CRIM1
Gene Name
cysteine rich transmembrane BMP regulator 1
There are no transcripts associated with this gene.

Gene
FEZ2
Gene Name
fasciculation and elongation protein zeta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042548.1 Intron NP_001036013.1
NM_005102.2 Intron NP_005093.2
XM_006712153.2 Intron XP_006712216.1

View Full Product Details