Product Details
- SNP ID
-
rs35549084
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:80415678 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAGCTCGGCTGAAGGCCCACGTC[A/G]TAGACCGGGACACCGAGGCGTGGCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ENDOV
PubMed Links
Gene Details
- Gene
- ENDOV
- Gene Name
- endonuclease V
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164637.2 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
NP_001158109.1 |
NM_001164638.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
NP_001158110.1 |
NM_173627.4 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
NP_775898.2 |
XM_005257244.3 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_005257301.1 |
XM_005257246.3 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_005257303.1 |
XM_006721837.3 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_006721900.1 |
XM_011524655.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522957.1 |
XM_011524656.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522958.1 |
XM_011524657.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522959.1 |
XM_011524658.2 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522960.1 |
XM_011524660.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522962.1 |
XM_011524661.2 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522963.1 |
XM_011524662.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522964.1 |
XM_011524663.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522965.1 |
XM_011524664.2 |
163 |
UTR 5 |
|
|
XP_011522966.1 |
XM_011524666.1 |
163 |
UTR 5 |
|
|
XP_011522968.1 |
XM_011524667.2 |
163 |
UTR 5 |
|
|
XP_011522969.1 |
XM_011524669.2 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522971.1 |
XM_011524670.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522972.1 |
XM_011524671.2 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_011522973.1 |
XM_011524672.1 |
163 |
UTR 5 |
|
|
XP_011522974.1 |
XM_011524673.1 |
163 |
UTR 5 |
|
|
XP_011522975.1 |
XM_011524674.1 |
163 |
UTR 5 |
|
|
XP_011522976.1 |
XM_011524675.1 |
163 |
UTR 5 |
|
|
XP_011522977.1 |
XM_011524676.1 |
163 |
UTR 5 |
|
|
XP_011522978.1 |
XM_011524677.1 |
163 |
UTR 5 |
|
|
XP_011522979.1 |
XM_011524678.2 |
163 |
Intron |
|
|
XP_011522980.1 |
XM_017024508.1 |
163 |
UTR 5 |
|
|
XP_016879997.1 |
XM_017024509.1 |
163 |
UTR 5 |
|
|
XP_016879998.1 |
XM_017024510.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_016879999.1 |
XM_017024511.1 |
163 |
Missense Mutation |
ATA,GTA |
I29V |
XP_016880000.1 |
XM_017024512.1 |
163 |
UTR 5 |
|
|
XP_016880001.1 |
XM_017024513.1 |
163 |
Intron |
|
|
XP_016880002.1 |
XM_017024514.1 |
163 |
UTR 5 |
|
|
XP_016880003.1 |
XM_017024515.1 |
163 |
UTR 5 |
|
|
XP_016880004.1 |
XM_017024516.1 |
163 |
Intron |
|
|
XP_016880005.1 |
XM_017024517.1 |
163 |
UTR 5 |
|
|
XP_016880006.1 |
XM_017024518.1 |
163 |
UTR 5 |
|
|
XP_016880007.1 |
XM_017024519.1 |
163 |
UTR 5 |
|
|
XP_016880008.1 |
XM_017024520.1 |
163 |
UTR 5 |
|
|
XP_016880009.1 |
- Gene
- LOC100294362
- Gene Name
- uncharacterized LOC100294362
There are no transcripts associated with this gene.
- Gene
- MIR4730
- Gene Name
- microRNA 4730
There are no transcripts associated with this gene.
View Full Product Details