Product Details

SNP ID
rs7246969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9115264 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGATTTCAACGTTTTTGCAGAAG[A/G]ACAGCTGCAATACCATCAGACTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR7G1 PubMed Links

Gene Details

Gene
OR7G1
Gene Name
olfactory receptor family 7 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005192.2 500 Missense Mutation TCC,TTC S167F NP_001005192.2

View Full Product Details