Product Details

SNP ID
rs58686906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79000932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGATAGACAGGTGGAGCTGTACC[G/T]GGGTCCTCATTTGGCAGCATCACTT
Phenotype
MIM: 605849
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DMGDH PubMed Links

Gene Details

Gene
DMGDH
Gene Name
dimethylglycine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013391.3 Intron NP_037523.2
XM_006714597.2 Intron XP_006714660.1
XM_011543354.2 Intron XP_011541656.1
XM_011543355.2 Intron XP_011541657.1

View Full Product Details