Product Details
- SNP ID
-
rs11558054
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:150692506 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGAAGCTGGGGAAATAGAGAGATT[G/T]TCATTATTGACACACCAGATATGTT
- Phenotype
-
MIM: 608085
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GIMAP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1860871] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GIMAP2
- Gene Name
- GTPase, IMAP family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015660.2 |
308 |
Missense Mutation |
GTC,TTC |
V74F |
NP_056475.1 |
View Full Product Details