Product Details

SNP ID
rs72785870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:30156496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATCTTTTTGTTCAGGATCAGCT[A/G]TTTCACACATCGAGTACTACAAAGC
Phenotype
MIM: 609726
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YPEL5 PubMed Links

Gene Details

Gene
YPEL5
Gene Name
yippee like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127399.1 Intron NP_001120871.1
NM_001127400.1 Intron NP_001120872.1
NM_001127401.1 Intron NP_001120873.1
NM_016061.2 Intron NP_057145.1
XM_017004318.1 Intron XP_016859807.1
XM_017004319.1 Intron XP_016859808.1
XM_017004320.1 Intron XP_016859809.1
XM_017004321.1 Intron XP_016859810.1
XM_017004322.1 Intron XP_016859811.1

View Full Product Details