Product Details

SNP ID
rs16860239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112924004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTGTTAACTACATAAGTCTCTG[C/T]GTGGAGATAATGGCTCTGTGCAAGT
Phenotype
MIM: 607546
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD200R1 PubMed Links

Gene Details

Gene
CD200R1
Gene Name
CD200 receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138806.3 Intron NP_620161.1
NM_138939.2 Intron NP_620385.1
NM_138940.2 Intron NP_620386.1
NM_170780.2 Intron NP_740750.1

View Full Product Details