Product Details

SNP ID
rs114780658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24119415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAATCTACCTTCCCCATGGCTC[A/T]GCTGCACCCTGTCCTTCCAGTGGCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM228B PubMed Links

Gene Details

Gene
FAM228B
Gene Name
family with sequence similarity 228 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145710.1 Intron NP_001139182.1
NM_001291328.1 Intron NP_001278257.1
Gene
PFN4
Gene Name
profilin family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199346.2 Intron NP_955378.1
XM_005264292.2 Intron XP_005264349.1
XM_006712007.3 Intron XP_006712070.1
XM_011532823.2 Intron XP_011531125.1

View Full Product Details