Product Details

SNP ID
rs139671261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:181539032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAGTTCAGAGAAGAGAGCTTTC[A/G]TTGTAATATTAGATTTATGTTTACT
Phenotype
MIM: 608381 MIM: 192975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERKL PubMed Links

Gene Details

Gene
CERKL
Gene Name
ceramide kinase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030311.2 Intron NP_001025482.1
NM_001030312.2 Intron NP_001025483.1
NM_001030313.2 Intron NP_001025484.1
NM_001160277.1 Intron NP_001153749.1
NM_201548.4 Intron NP_963842.1
Gene
ITGA4
Gene Name
integrin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000885.5 Intron NP_000876.3
NM_001316312.1 Intron NP_001303241.1

View Full Product Details