Product Details
- SNP ID
-
rs61745544
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:128051174 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACTGCTCATAGAGGTCATGATGG[A/T]TGCACCAATCCATGCAGAGAAGCAT
- Phenotype
-
MIM: 300487
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ACTRT1
PubMed Links
Gene Details
- Gene
- ACTRT1
- Gene Name
- actin related protein T1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138289.3 |
1230 |
Missense Mutation |
ACC,TCC |
T345S |
NP_612146.1 |
View Full Product Details