Product Details

SNP ID
rs184370918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18138018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCTGACGGCGGCCACAGTGGC[C/T]GGCGTACATGTGAAGCAGCAGTGGG
Phenotype
MIM: 614771
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KAT14 PubMed Links

Gene Details

Gene
KAT14
Gene Name
lysine acetyltransferase 14
There are no transcripts associated with this gene.

Gene
PET117
Gene Name
PET117 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164811.1 164 Silent Mutation GCC,GCT A21A NP_001158283.1

View Full Product Details