Product Details

SNP ID
rs80088378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:29193171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGAAGGAGCCATTGCCTCTCT[C/G]TCCTCCACGGTCTTAGGGATCCCAA
Phenotype
MIM: 105590
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALK PubMed Links

Gene Details

Gene
ALK
Gene Name
anaplastic lymphoma receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004304.4 5868 UTR 3 NP_004295.2
Gene
CLIP4
Gene Name
CAP-Gly domain containing linker protein family member 4
There are no transcripts associated with this gene.

View Full Product Details