Product Details

SNP ID
rs34130630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35468653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTTCCTCAGGGCCAGGTAGCC[A/G]GTGACGACGTCAGAGGGCAGCCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INO80C PubMed Links

Gene Details

Gene
INO80C
Gene Name
INO80 complex subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098817.1 762 Silent Mutation ACC,ACT T215T NP_001092287.1
NM_001308064.1 762 Silent Mutation ACC,ACT T124T NP_001294993.1
NM_194281.3 762 Silent Mutation ACC,ACT T179T NP_919257.2
XM_005258196.4 762 Silent Mutation ACC,ACT T124T XP_005258253.1
XM_005258197.4 762 Silent Mutation ACC,ACT T124T XP_005258254.1
XM_011525812.2 762 Silent Mutation ACC,ACT T153T XP_011524114.1
XM_017025549.1 762 Silent Mutation ACC,ACT T124T XP_016881038.1

View Full Product Details