Product Details

SNP ID
rs11551368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31672186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCTCTGAGAGTTATAACTTAA[C/G]ACAATAAAATAATTAAAAATAGTGT
Phenotype
MIM: 615256
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AMN1 PubMed Links

Gene Details

Gene
AMN1
Gene Name
antagonist of mitotic exit network 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113402.1 4354 UTR 3 NP_001106873.1
NM_001278411.1 4354 UTR 3 NP_001265340.1
NM_001278412.1 4354 UTR 3 NP_001265341.1
XM_006719054.3 4354 UTR 3 XP_006719117.1
XM_011520604.2 4354 UTR 3 XP_011518906.1
XM_017018963.1 4354 UTR 3 XP_016874452.1
XM_017018964.1 4354 UTR 3 XP_016874453.1
XM_017018965.1 4354 UTR 3 XP_016874454.1
Gene
ETFBKMT
Gene Name
electron transfer flavoprotein beta subunit lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135863.1 4354 Intron NP_001129335.1
NM_001135864.1 4354 Intron NP_001129336.1
NM_173802.3 4354 Intron NP_776163.1

View Full Product Details