Product Details

SNP ID
rs61738638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17203007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCTGGATGGCTTGCGTGGTCCA[G/T]TTGAGCGAGCCAGTGATGAGCACCC
Phenotype
MIM: 607273 MIM: 614960
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FLCN PubMed Links

Gene Details

Gene
FLCN
Gene Name
folliculin
There are no transcripts associated with this gene.

Gene
PLD6
Gene Name
phospholipase D family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178836.3 565 Missense Mutation AAA,AAC K173N NP_849158.2
XM_017024310.1 565 Missense Mutation AAT,ACT N172T XP_016879799.1

View Full Product Details