Product Details

SNP ID
rs3014830
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:153437166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCAGAATAATTAACCTTCTTATC[C/T]TTATTATCATCTTTTTTCTCAAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100A7L2 PubMed Links

Gene Details

Gene
S100A7L2
Gene Name
S100 calcium binding protein A7 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045479.1 309 Silent Mutation NP_001038944.2

View Full Product Details