Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042632.2 | 607 | UTR 3 | NP_001036097.1 | ||
NM_001042633.2 | 607 | UTR 3 | NP_001036098.1 | ||
NM_033421.3 | 607 | Missense Mutation | TTA,TTT | L175F | NP_219489.1 |
NM_152897.2 | 607 | Missense Mutation | TTA,TTT | L175F | NP_690857.1 |
XM_005260608.4 | 607 | Missense Mutation | TTA,TTT | L77F | XP_005260665.1 |
XM_011529095.2 | 607 | Missense Mutation | TTA,TTT | L27F | XP_011527397.1 |
XM_011529096.2 | 607 | Intron | XP_011527398.1 | ||
XM_011529098.2 | 607 | UTR 3 | XP_011527400.1 | ||
XM_011529099.2 | 607 | Intron | XP_011527401.1 | ||
XM_017028120.1 | 607 | Intron | XP_016883609.1 |