Product Details
- SNP ID
-
rs35294030
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:169779040 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAGCGAAAAAGTCATCTCAAACGA[A/G]AAAGAAGAAGAAGGCTTTCAACTCC
- Phenotype
-
MIM: 608534
MIM: 606042
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTRT3
PubMed Links
Gene Details
- Gene
- ACTRT3
- Gene Name
- actin related protein T3
There are no transcripts associated with this gene.
- Gene
- MYNN
- Gene Name
- myoneurin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001185118.1 |
1726 |
Missense Mutation |
AAA,AGA |
K180R |
NP_001172047.1 |
NM_001185119.1 |
1726 |
Missense Mutation |
AAA,AGA |
K180R |
NP_001172048.1 |
NM_018657.4 |
1726 |
Missense Mutation |
AAA,AGA |
K180R |
NP_061127.1 |
XM_005247621.4 |
1726 |
Missense Mutation |
AAA,AGA |
K180R |
XP_005247678.1 |
XM_005247622.3 |
1726 |
Missense Mutation |
AAA,AGA |
K98R |
XP_005247679.1 |
XM_005247624.3 |
1726 |
Missense Mutation |
AAA,AGA |
K66R |
XP_005247681.1 |
XM_011512987.1 |
1726 |
Missense Mutation |
AAA,AGA |
K180R |
XP_011511289.1 |
XM_017006864.1 |
1726 |
Missense Mutation |
AAA,AGA |
K180R |
XP_016862353.1 |
XM_017006865.1 |
1726 |
Missense Mutation |
AAA,AGA |
K66R |
XP_016862354.1 |
XM_017006866.1 |
1726 |
Missense Mutation |
AAA,AGA |
K66R |
XP_016862355.1 |
XM_017006867.1 |
1726 |
UTR 5 |
|
|
XP_016862356.1 |
XM_017006868.1 |
1726 |
UTR 5 |
|
|
XP_016862357.1 |
View Full Product Details