Product Details

SNP ID
rs61742567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4803522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATGATGGGGTTGAGCATGGGAG[G/T]TATCAGTAGATAGAGATTAGCAAAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR52R1 PubMed Links

Gene Details

Gene
OR52R1
Gene Name
olfactory receptor family 52 subfamily R member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005177.3 859 Missense Mutation ACT,CCT T287P NP_001005177.3

View Full Product Details