Product Details

SNP ID
rs116538588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33697541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAAGGGCTTCCTTTCTGGGAAA[A/G]CTAGAGGAGATTCCCAAACCGTAAG
Phenotype
MIM: 147267 MIM: 614461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITPR3 PubMed Links

Gene Details

Gene
ITPR3
Gene Name
inositol 1,4,5-trisphosphate receptor type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002224.3 558 Intron NP_002215.2
XM_011514577.2 558 Intron XP_011512879.1
XM_017010832.1 558 Intron XP_016866321.1
Gene
MIR3934
Gene Name
microRNA 3934
There are no transcripts associated with this gene.

Gene
UQCC2
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032340.3 558 UTR 3 NP_115716.1

View Full Product Details