Product Details

SNP ID
rs17104322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:146223371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCAAATTTTTGTTTTTTGTTTG[C/T]GCAATATGTAAATGTTATTTCTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM27 PubMed Links

Gene Details

Gene
RBM27
Gene Name
RNA binding motif protein 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018989.1 Intron NP_061862.1
XM_005268466.1 Intron XP_005268523.1
XM_005268467.1 Intron XP_005268524.1
XM_005268468.1 Intron XP_005268525.1
XM_006714787.1 Intron XP_006714850.1
XM_011537661.1 Intron XP_011535963.1
XM_011537662.1 Intron XP_011535964.1
XM_017009587.1 Intron XP_016865076.1

View Full Product Details