Product Details

SNP ID
rs923283257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30164357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATGGAAAGAAAGCTCCAATGCC[A/G]AGTGGGATCTTCTGCAGATTATGGG
Phenotype
MIM: 605701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM10 PubMed Links

Gene Details

Gene
TRIM10
Gene Name
tripartite motif containing 10
There are no transcripts associated with this gene.

Gene
TRIM15
Gene Name
tripartite motif containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033229.2 Intron NP_150232.2

View Full Product Details