Product Details
- SNP ID
-
rs12625959
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23444023 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACGAGCAATTCTTCCTGCCCCCT[C/G]CAAAGCAAGAAGCTGAGAAAGGTGT
- Phenotype
-
MIM: 609731
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CST11
PubMed Links
Gene Details
- Gene
- CST11
- Gene Name
- cystatin 11
There are no transcripts associated with this gene.
- Gene
- CSTL1
- Gene Name
- cystatin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138283.1 |
555 |
Silent Mutation |
CTC,CTG |
L103L |
NP_612140.1 |
View Full Product Details