Product Details

SNP ID
rs564278107
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:186446312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTTCCATTTTTTTTTGCCAGC[A/G]TAAGCAATATTGTAATGAATATATT
Phenotype
MIM: 171490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724919 PubMed Links

Gene Details

Gene
LOC102724919
Gene Name
uncharacterized LOC102724919
There are no transcripts associated with this gene.

Gene
PDC
Gene Name
phosducin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002597.4 Intron NP_002588.3
NM_022576.3 Intron NP_072098.1
XM_011509603.2 Intron XP_011507905.1

View Full Product Details