Product Details

SNP ID
rs61743326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48498288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTGGTCCCCGTTCTCCAGCAC[A/T]GTCACCCCGTTCACTGGGAGGCTCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LMTK3 PubMed Links

Gene Details

Gene
LMTK3
Gene Name
lemur tyrosine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080434.1 3199 Silent Mutation ACA,ACT T956T NP_001073903.1
XM_011526411.2 3199 Silent Mutation ACA,ACT T953T XP_011524713.1
XM_011526412.2 3199 Silent Mutation ACA,ACT T942T XP_011524714.1

View Full Product Details