Product Details

SNP ID
rs550615736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222572222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAACTCTTGCAAGGAAAAAATC[C/T]GGGGCAAATATGAAAATAAGGGCCC
Phenotype
MIM: 609690 MIM: 612827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FARSB PubMed Links

Gene Details

Gene
FARSB
Gene Name
phenylalanyl-tRNA synthetase beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005687.4 Intron NP_005678.3
XM_006712169.2 Intron XP_006712232.1
XM_011510466.2 Intron XP_011508768.1
XM_017003110.1 Intron XP_016858599.1
Gene
SGPP2
Gene Name
sphingosine-1-phosphate phosphatase 2
There are no transcripts associated with this gene.

View Full Product Details