Product Details
- SNP ID
-
rs61747070
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:65384280 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCGAGATGAGATCAGGAGCCTCT[A/G]GAGGCAGGGGGTCCTCAGGCTCCAG
- Phenotype
-
MIM: 604184
MIM: 616810
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IGDCC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2292933] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IGDCC3
- Gene Name
- immunoglobulin superfamily DCC subclass member 3
There are no transcripts associated with this gene.
- Gene
- IGDCC4
- Gene Name
- immunoglobulin superfamily DCC subclass member 4
View Full Product Details