Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001272065.1 | 1104 | Intron | NP_001258994.1 | ||
NM_001272066.1 | 1104 | UTR 3 | NP_001258995.1 | ||
NM_001272067.1 | 1104 | UTR 3 | NP_001258996.1 | ||
NM_001272068.1 | 1104 | UTR 3 | NP_001258997.1 | ||
NM_001272082.1 | 1104 | UTR 3 | NP_001259011.1 | ||
NM_001272083.1 | 1104 | UTR 3 | NP_001259012.1 | ||
NM_016479.4 | 1104 | Intron | NP_057563.3 | ||
XM_005265205.1 | 1104 | UTR 3 | XP_005265262.1 | ||
XM_006713188.2 | 1104 | UTR 3 | XP_006713251.1 | ||
XM_011533796.1 | 1104 | UTR 3 | XP_011532098.1 |