Product Details

SNP ID
rs112933459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52420116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGTCCTAAGTGAGAAGAGGGAG[A/G]AACATCTTGAGAAAATAGGAAAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHF7 PubMed Links

Gene Details

Gene
PHF7
Gene Name
PHD finger protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278221.2 Intron NP_001265150.1
NM_001321126.1 Intron NP_001308055.1
NM_001321127.1 Intron NP_001308056.1
NM_016483.6 Intron NP_057567.3
XM_011533826.2 Intron XP_011532128.1

View Full Product Details