Product Details
- SNP ID
-
rs61743588
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
22
- Location
-
Chr.1:153438206 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACTCACACAGCCACTGAGGAAGT[C/T]GGGGAAGTTCTCCTTCATCAAGTTC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
S100A7L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147282626] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- S100A7L2
- Gene Name
- S100 calcium binding protein A7 like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001045479.1 |
235 |
Missense Mutation |
AAC,GAC |
N53D |
NP_001038944.2 |
View Full Product Details