Product Details

SNP ID
rs62131196
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1032475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACCAGGTGAGGGGCTGGTGGA[G/T]CGGAGCAGGGATGGTGCTGGGGGCC
Phenotype
MIM: 605414 MIM: 602373
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ABCA7 PubMed Links

Gene Details

Gene
ABCA7
Gene Name
ATP binding cassette subfamily A member 7
There are no transcripts associated with this gene.

Gene
CNN2
Gene Name
calponin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303499.1 Intron NP_001290428.1
NM_001303501.1 Intron NP_001290430.1
NM_004368.3 Intron NP_004359.1
NM_201277.2 Intron NP_958434.1

View Full Product Details