Product Details

SNP ID
rs113067724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77428509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACAAACAATACAAACCTCCAAT[C/G]AGCCTCTGTACAGGGTTCTTTTTGC
Phenotype
MIM: 613401
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOXRED1 PubMed Links

Gene Details

Gene
NOXRED1
Gene Name
NADP dependent oxidoreductase domain containing 1
There are no transcripts associated with this gene.

Gene
VIPAS39
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193314.1 Intron NP_001180243.1
NM_001193315.1 Intron NP_001180244.1
NM_001193316.1 Intron NP_001180245.1
NM_001193317.1 Intron NP_001180246.1
NM_022067.3 Intron NP_071350.2
XM_011537066.1 Intron XP_011535368.1
XM_017021579.1 Intron XP_016877068.1
XM_017021580.1 Intron XP_016877069.1
XM_017021581.1 Intron XP_016877070.1

View Full Product Details