Product Details
- SNP ID
-
rs4459050
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
39
- Location
-
Chr.1:1918091 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACAGGCTGTAGGCAGGAGGAGCCA[C/T]GGAGTCCAGGTCCAGCTCCCCAAAG
- Phenotype
-
MIM: 610171
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CALML6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28640257] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CALML6
- Gene Name
- calmodulin like 6
- Gene
- CFAP74
- Gene Name
- cilia and flagella associated protein 74
There are no transcripts associated with this gene.
- Gene
- TMEM52
- Gene Name
- transmembrane protein 52
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178545.3 |
457 |
Missense Mutation |
ATG,GTG |
M141V |
NP_848640.1 |
View Full Product Details