Product Details

SNP ID
rs66846694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55746115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTATTTGTGAAATAAATGACAC[A/G]TTCATATTCAAAGTAGGAAACTAGT
Phenotype
MIM: 605281
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX4 PubMed Links

Gene Details

Gene
DDX4
Gene Name
DEAD-box helicase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142549.1 Intron NP_001136021.1
NM_001166533.1 Intron NP_001160005.1
NM_001166534.1 Intron NP_001160006.1
NM_024415.2 Intron NP_077726.1
XM_011543495.2 Intron XP_011541797.1
XM_011543497.2 Intron XP_011541799.1

View Full Product Details