Product Details

SNP ID
rs7283603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34180828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCTGCTCAACTCCCTCTTCGTT[A/T]TGGTGTGGGGCGGGGGTGGGGGTGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00310 PubMed Links

Gene Details

Gene
LINC00310
Gene Name
long intergenic non-protein coding RNA 310
There are no transcripts associated with this gene.

View Full Product Details