Product Details

SNP ID
rs12269847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102399012 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAGCTGCACAATCAAAATGGTCA[C/G]TGTTCTGTGTAAAGTTCCTTAAGTA
Phenotype
MIM: 606356
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM123 PubMed Links

Gene Details

Gene
TMEM123
Gene Name
transmembrane protein 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052932.2 Intron NP_443164.2

View Full Product Details