Product Details

SNP ID
rs61744556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:3599597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACACCGAGGGCGACCCGGAGAAG[C/G]CCGAGGACGACGAGGAGATCGACCT
Phenotype
MIM: 606197
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRX1 PubMed Links

Gene Details

Gene
IRX1
Gene Name
iroquois homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024337.3 701 Missense Mutation CCC,GCC P217A NP_077313.3
XM_017009845.1 701 Missense Mutation CCC,GCC P217A XP_016865334.1

View Full Product Details