Product Details

SNP ID
rs57405643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:168010657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGTACCATTACCCACTGGTAC[C/G]TATCCCAAATACATATATCTCTCCT
Phenotype
MIM: 606805
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GOLIM4 PubMed Links

Gene Details

Gene
GOLIM4
Gene Name
golgi integral membrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308155.1 Intron NP_001295084.1
NM_014498.4 Intron NP_055313.1
XM_005247364.2 Intron XP_005247421.1
XM_005247365.2 Intron XP_005247422.1
XM_017006183.1 Intron XP_016861672.1

View Full Product Details