Product Details

SNP ID
rs148762006
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39995624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGGCAGGGCCCGGGGCAGGCC[A/G]CCCTGAGCACCCTCTGGAAAATGGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FAM83F PubMed Links

Gene Details

Gene
FAM83F
Gene Name
family with sequence similarity 83 member F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138435.2 Intron NP_612444.2

View Full Product Details