Product Details

SNP ID
rs77282321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125721607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAAGATATACATACAATATCA[A/G]TTGCATATGCTTCTTTTAAGTCTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDRF1 PubMed Links

Gene Details

Gene
EDRF1
Gene Name
erythroid differentiation regulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202438.1 Intron NP_001189367.1
NM_015608.2 Intron NP_056423.2
XM_011539622.1 Intron XP_011537924.1
XM_017016066.1 Intron XP_016871555.1
Gene
FLJ37035
Gene Name
uncharacterized LOC399821
There are no transcripts associated with this gene.

View Full Product Details