Product Details

SNP ID
rs9478409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:150064750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAAAAGTGACAACTCTTGTCCA[C/T]GCTCCAAATCTGAGGAGATGGCTCC
Phenotype
MIM: 605699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ULBP3 PubMed Links

Gene Details

Gene
ULBP3
Gene Name
UL16 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024518.2 Intron NP_078794.1

View Full Product Details