Product Details

SNP ID
rs148165748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:56059162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTATGAGCAGGTAGCAATTCT[C/T]ACCTGGGGGACCAGGAGGACCTTGT
Phenotype
MIM: 610002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL21A1 PubMed Links

Gene Details

Gene
COL21A1
Gene Name
collagen type XXI alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318751.1 Intron NP_001305680.1
NM_001318752.1 Intron NP_001305681.1
NM_001318753.1 Intron NP_001305682.1
NM_001318754.1 Intron NP_001305683.1
NM_030820.3 Intron NP_110447.2
XM_006715223.1 Intron XP_006715286.1
XM_011514924.2 Intron XP_011513226.1
XM_011514925.2 Intron XP_011513227.1
XM_011514926.1 Intron XP_011513228.1
XM_011514927.1 Intron XP_011513229.1

View Full Product Details