Product Details

SNP ID
rs148736034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73443170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCAGGTCCCCGCCGGCAGTGCG[A/T]GTGGTGAGGCAGGACATGGCGGAGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC107984728 PubMed Links

Gene Details

Gene
LOC107984728
Gene Name
serine-aspartate repeat-containing protein I-like
There are no transcripts associated with this gene.

Gene
REC114
Gene Name
REC114 meiotic recombination protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042367.1 13 UTR 5 NP_001035826.1
XM_017022083.1 13 Intron XP_016877572.1

View Full Product Details