Product Details

SNP ID
rs3796158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195069693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCCCCCAGATCTGGAGGCCCC[A/G]GGGGCAAGGCACGGGGAGCGCCTAG
Phenotype
MIM: 614552
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XXYLT1 PubMed Links

Gene Details

Gene
XXYLT1
Gene Name
xyloside xylosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308069.1 1312 UTR 3 NP_001294998.1
NM_152531.4 1312 UTR 3 NP_689744.3
XM_005269286.4 1312 UTR 3 XP_005269343.1
XM_011512448.2 1312 UTR 3 XP_011510750.1
XM_017005749.1 1312 Intron XP_016861238.1
XM_017005750.1 1312 UTR 3 XP_016861239.1

View Full Product Details