Product Details

SNP ID
rs73058308
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55616877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGACTGATCAGACTCTTCCCC[C/G]CTCCTCGCTGTTGCCCCATCCTTCA
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF784 PubMed Links

Gene Details

Gene
ZNF784
Gene Name
zinc finger protein 784
There are no transcripts associated with this gene.

Gene
ZNF865
Gene Name
zinc finger protein 865
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195605.1 3285 UTR 3 NP_001182534.1

View Full Product Details