Product Details

SNP ID
rs35199358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142940366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTTCCAGACCAGAATCAAGAA[C/G]AGTTCGAAGAGCACTTTGTGGCCTC
Phenotype
MIM: 613883 MIM: 606679
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C7orf34 PubMed Links

Gene Details

Gene
C7orf34
Gene Name
chromosome 7 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178829.4 264 Missense Mutation CAG,GAG Q75E NP_849151.2
Gene
KEL
Gene Name
Kell blood group, metallo-endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000420.2 264 Intron NP_000411.1
XM_005249993.2 264 Intron XP_005250050.1
XM_005249994.3 264 Intron XP_005250051.1
Gene
TRPV5
Gene Name
transient receptor potential cation channel subfamily V member 5
There are no transcripts associated with this gene.

View Full Product Details