Product Details

SNP ID
hCV25940433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50723582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTTCTCTGCTGTTTTCTGTC[C/G]CTGTTCAGTCACTTCCAGAGCTTTC
Phenotype
MIM: 608684
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NIN PubMed Links

Gene Details

Gene
NIN
Gene Name
ninein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016350.4 6474 Intron NP_057434.4
NM_020921.3 6474 Missense Mutation CGA,GGA R2095G NP_065972.3
NM_182944.2 6474 Intron NP_891989.2
NM_182946.1 6474 Intron NP_891991.1

View Full Product Details