Product Details

SNP ID
rs142210491
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:441326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGGACTGATGCTGTTGTCTGCA[C/G]AGGACAGCGTGGCTCTCCAGGACTC
Phenotype
MIM: 615928
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
FAM101B PubMed Links

Gene Details

Gene
FAM101B
Gene Name
family with sequence similarity 101 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182705.2 2547 UTR 3 NP_874364.1

View Full Product Details